Assay Details
| Gene Symbol: SMARCD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751852.4 | XP_002751898.1 | 7 - 8 | 872 | 94 | |
| XM_002751854.3 | XP_002751900.1 | 8 - 9 | 1217 | 94 | ||
| XM_017975506.1 | XP_017830995.1 | 7 - 8 | 662 | 94 | ||
More Information
Set Membership: |
|

SMARCD3
Genomic Map