Assay Details
| Gene Symbol: NSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748344.4 | XP_002748390.2 | 3 - 4 | 245 | 89 | |
| XM_008997145.2 | XP_008995393.1 | 4 - 5 | 378 | 89 | ||
| XM_008997146.2 | XP_008995394.1 | 4 - 5 | 214 | 89 | ||
| XM_017972555.1 | XP_017828044.1 | 3 - 4 | 317 | 89 | ||
| XM_017972556.1 | XP_017828045.1 | 2 - 3 | 77 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008997146.2, XM_017972555.1, XM_017972556.1 transcripts.
Set Membership: |
|

NSRP1
Genomic Map