Assay Details
| Gene Symbol: VPS29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752999.4 | XP_002753045.1 | 3 - 4 | 353 | 97 | |
| XM_002753000.4 | XP_002753046.1 | 2 - 3 | 277 | 97 | ||
| XM_009004616.2 | XP_009002864.1 | 3 - 4 | 237 | 97 | ||
More Information
Set Membership: |
|

VPS29
Genomic Map