Assay Details
| Gene Symbol: TAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750519.4 | XP_002750565.1 | 4 - 5 | 511 | 86 | |
| XM_009000760.2 | XP_008999008.1 | 3 - 4 | 486 | 86 | ||
| XM_009000761.2 | XP_008999009.1 | 3 - 4 | 497 | 86 | ||
| XM_009000762.1 | XP_008999010.1 | 3 - 4 | 440 | 86 | ||
More Information
Set Membership: |
|

TAF12
Genomic Map