Assay Details
| Gene Symbol: SYNCRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746784.3 | XP_002746830.1 | 8 - 9 | 1529 | 86 | |
| XM_008994735.2 | XP_008992983.1 | 8 - 9 | 1535 | 86 | ||
| XM_008994736.2 | XP_008992984.1 | 8 - 9 | 1535 | 86 | ||
| XM_008994737.2 | XP_008992985.1 | 7 - 8 | 1375 | 86 | ||
| XM_008994738.2 | XP_008992986.1 | 7 - 8 | 1375 | 86 | ||
| XM_017971242.1 | XP_017826731.1 | 8 - 9 | 1128 | 86 | ||
More Information
Set Membership: |
|

SYNCRIP
Genomic Map