Assay Details
| Gene Symbol: GABRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763372.3 | XP_002763418.1 | 7 - 8 | 1020 | 75 | |
| XM_017968719.1 | XP_017824208.1 | 8 - 9 | 970 | 75 | ||
| XM_017968720.1 | XP_017824209.1 | 6 - 7 | 870 | 75 | ||
More Information
Set Membership: |
|

GABRA3
Genomic Map