Assay Details
| Gene Symbol: TUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750830.4 | XP_002750876.2 | 20 - 21 | 3793 | 89 | |
| XM_009001219.2 | XP_008999467.1 | 20 - 21 | 3793 | 89 | ||
| XM_009001220.2 | XP_008999468.1 | 20 - 21 | 3793 | 89 | ||
| XM_009001221.2 | XP_008999469.1 | 20 - 21 | 3793 | 89 | ||
| XM_009001222.2 | XP_008999470.1 | 20 - 21 | 3793 | 89 | ||
| XM_009001224.2 | XP_008999472.1 | 19 - 20 | 3580 | 89 | ||
| XM_009001225.2 | XP_008999473.1 | 19 - 20 | 3012 | 89 | ||
| XM_017974741.1 | XP_017830230.1 | 20 - 21 | 3618 | 89 | ||
More Information
Set Membership: |
|

TUT4
Genomic Map