Assay Details
| Gene Symbol: TMEFF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743142.4 | XP_002743188.1 | 9 - 10 | 1292 | 94 | |
| XM_009000279.1 | XP_008998527.1 | 9 - 10 | 1288 | 94 | ||
More Information
Set Membership: |
|

TMEFF1
Genomic Map