Assay Details
| Gene Symbol: SERINC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747577.4 | XP_002747623.2 | 9 - 10 | 1449 | 78 | |
| GenBank mRNA | EF215349.1 | - | 1 - 2 | 198 | 78 | |
More Information
Set Membership: |
|

SERINC3
Genomic Map