Assay Details
| Gene Symbol: SLC25A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971758.1 | XP_017827247.1 | 1 - 2 | 749 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC25A10| Gene Symbol: SLC25A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971758.1 | XP_017827247.1 | 1 - 2 | 749 | 118 | |
Set Membership: |
|