Assay Details
| Gene Symbol: MAT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744140.4 | XP_002744186.1 | 5 - 6 | 867 | 74 | |
| XM_002744141.4 | XP_002744187.1 | 5 - 6 | 1006 | 74 | ||
More Information
Set Membership: |
|

MAT2B
Genomic Map