Assay Details
| Gene Symbol: CHST15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756689.4 | XP_002756735.2 | 6 - 7 | 3223 | 75 | |
| XM_008978859.2 | XP_008977107.1 | 6 - 7 | 3140 | 75 | ||
| XM_008978860.2 | XP_008977108.1 | 6 - 7 | 1921 | 75 | ||
More Information
Set Membership: |
|

CHST15
Genomic Map