Assay Details
| Gene Symbol: TRMT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971949.1 | XP_017827438.1 | 6 - 7 | 772 | 64 | |
| XM_017971950.1 | XP_017827439.1 | 6 - 7 | 772 | 64 | ||
More Information
Set Membership: |
|

TRMT6
Genomic Map