Assay Details
| Gene Symbol: KLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745918.3 | XP_002745964.2 | 4 - 5 | 940 | 78 | |
| XM_008993489.2 | XP_008991737.1 | 4 - 5 | 929 | 78 | ||
| XM_008993490.2 | XP_008991738.1 | 4 - 5 | 821 | 78 | ||
More Information
Set Membership: |
|

KLF3
Genomic Map