Assay Details
| Gene Symbol: IMPAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758924.4 | XP_002758970.1 | 4 - 5 | 1086 | 98 | |
| XM_008982676.2 | XP_008980924.1 | 3 - 4 | 990 | 98 | ||
More Information
Set Membership: |
|

IMPAD1
Genomic Map