Assay Details
| Gene Symbol: FBXL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009460.2 | XP_009007708.1 | 8 - 9 | 2160 | 91 | |
| XM_017978872.1 | XP_017834361.1 | 8 - 9 | 2031 | 91 | ||
More Information
Set Membership: |
|

FBXL19
Genomic Map