Assay Details
| Gene Symbol: MOV10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751234.3 | XP_002751280.1 | 17 - 18 | 3094 | 68 | |
| XM_009001901.1 | XP_009000149.1 | 17 - 18 | 3078 | 68 | ||
| XM_009001902.2 | XP_009000150.1 | 16 - 17 | 2568 | 68 | ||
| XM_017975028.1 | XP_017830517.1 | 16 - 17 | 2902 | 68 | ||
More Information
Set Membership: |
|

MOV10
Genomic Map