Assay Details
| Gene Symbol: TUT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742884.4 | XP_002742930.2 | 4 - 5 | 1092 | 92 | |
| XM_008996078.2 | XP_008994326.1 | 4 - 5 | 1055 | 92 | ||
| XM_008996088.2 | XP_008994336.1 | 4 - 5 | 1092 | 92 | ||
| XM_008996097.2 | XP_008994345.1 | 4 - 5 | 1092 | 92 | ||
| XM_017973193.1 | XP_017828682.1 | 4 - 5 | 1092 | 92 | ||
| XM_017973197.1 | XP_017828686.1 | 4 - 5 | 1092 | 92 | ||
| XM_017973209.1 | XP_017828698.1 | 4 - 5 | 1092 | 92 | ||
| XM_017973216.1 | XP_017828705.1 | 4 - 5 | 1092 | 92 | ||
More Information
Set Membership: |
|

TUT7
Genomic Map