Assay Details
| Gene Symbol: ALG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754378.4 | XP_002754424.2 | 5 - 6 | 640 | 81 | |
| XM_003734099.3 | XP_003734147.1 | 5 - 6 | 642 | 81 | ||
| XM_009006762.2 | XP_009005010.1 | 5 - 6 | 644 | 81 | ||
| XM_009006764.2 | XP_009005012.1 | 5 - 6 | 648 | 81 | ||
| XM_009006768.2 | XP_009005016.1 | 5 - 6 | 952 | 81 | ||
| XM_009006769.2 | XP_009005017.1 | 5 - 6 | 952 | 81 | ||
| XM_017977538.1 | XP_017833027.1 | 5 - 6 | 651 | 81 | ||
More Information
Set Membership: |
|

ALG9
Genomic Map