Assay Details
| Gene Symbol: LOC100398578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762895.3 | XP_002762941.1 | 3 - 4 | 1243 | 65 | |
| XM_002762896.2 | XP_002762942.1 | 3 - 4 | 1213 | 65 | ||
| XM_008989313.2 | XP_008987561.1 | 3 - 4 | 1241 | 65 | ||
| XM_008989314.1 | XP_008987562.1 | 3 - 4 | 1213 | 65 | ||
| XM_017968728.1 | XP_017824217.1 | 3 - 4 | 1241 | 65 | ||
| XM_017968729.1 | XP_017824218.1 | 3 - 4 | 1159 | 65 | ||
| XM_017968730.1 | XP_017824219.1 | 3 - 4 | 1241 | 65 | ||
More Information
Set Membership: |
|

LOC100398578
Genomic Map