Assay Details
| Gene Symbol: TBCEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754514.3 | XP_002754560.1 | 2 - 3 | 334 | 62 | |
| XM_009007002.2 | XP_009005250.1 | 2 - 3 | 4510 | 62 | ||
More Information
Set Membership: |
|

TBCEL
Genomic Map