Assay Details
| Gene Symbol: LOC100387916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758966.3 | XP_002759012.1 | 16 - 17 | 2349 | 102 | |
| XM_008982729.2 | XP_008980977.1 | 17 - 18 | 1885 | 102 | ||
| XM_008982730.2 | XP_008980978.1 | 16 - 17 | 1540 | 102 | ||
| XM_008982735.2 | XP_008980983.1 | 15 - 16 | 1777 | 102 | ||
| XM_008982736.2 | XP_008980984.1 | 14 - 15 | 1432 | 102 | ||
| XM_017965376.1 | XP_017820865.1 | 17 - 18 | 1599 | 102 | ||
| XM_017965377.1 | XP_017820866.1 | 16 - 17 | 1528 | 102 | ||
| XM_017965378.1 | XP_017820867.1 | 16 - 17 | 1590 | 102 | ||
| XM_017965379.1 | XP_017820868.1 | 13 - 14 | 1147 | 102 | ||
More Information
Set Membership: |
|

LOC100387916
Genomic Map