Assay Details
| Gene Symbol: LOC100410987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756516.3 | XP_002756562.1 | 3 - 4 | 1409 | 109 | |
| XM_017979229.1 | XP_017834718.1 | 4 - 5 | 512 | 109 | ||
More Information
Set Membership: |
|

LOC100410987
Genomic Map