Assay Details
| Gene Symbol: MINOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750374.4 | XP_002750420.1 | 1 - 2 | 153 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MINOS1| Gene Symbol: MINOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750374.4 | XP_002750420.1 | 1 - 2 | 153 | 114 | |
Set Membership: |
|