Assay Details
| Gene Symbol: PLA2G4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733934.2 | XP_003733982.1 | 14 - 15 | 1594 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G4D| Gene Symbol: PLA2G4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733934.2 | XP_003733982.1 | 14 - 15 | 1594 | 59 | |
Set Membership: |
|