Assay Details
| Gene Symbol: WASHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752907.2 | XP_002752953.1 | 5 - 6 | 596 | 113 | |
| XM_009004489.1 | XP_009002737.1 | 5 - 6 | 593 | 113 | ||
More Information
Set Membership: |
|

WASHC3
Genomic Map