Assay Details
| Gene Symbol: PQBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762843.4 | XP_002762889.1 | 2 - 3 | 242 | 76 | |
| XM_002762844.3 | XP_002762890.1 | 2 - 3 | 180 | 76 | ||
| XM_008989250.1 | XP_008987498.1 | 2 - 3 | 250 | 76 | ||
| XM_008989251.1 | XP_008987499.1 | 2 - 3 | 302 | 76 | ||
| XM_008989252.1 | XP_008987500.1 | 2 - 3 | 207 | 76 | ||
| XM_008989253.1 | XP_008987501.1 | 2 - 3 | 255 | 76 | ||
| XM_008989254.1 | XP_008987502.1 | 2 - 3 | 266 | 76 | ||
| XM_008989255.2 | XP_008987503.1 | 2 - 3 | 180 | 76 | ||
| XM_017968471.1 | XP_017823960.1 | 1 - 2 | 454 | 76 | ||
More Information
Set Membership: |
|

PQBP1
Genomic Map