Assay Details
| Gene Symbol: CDYL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746254.4 | XP_002746300.1 | 4 - 5 | 1407 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDYL| Gene Symbol: CDYL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746254.4 | XP_002746300.1 | 4 - 5 | 1407 | 101 | |
Set Membership: |
|