Assay Details
| Gene Symbol: LEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745507.3 | XP_002745553.1 | 8 - 9 | 2204 | 100 | |
| XM_003732454.2 | XP_003732502.1 | 9 - 10 | 2288 | 100 | ||
| XM_008992872.2 | XP_008991120.1 | 9 - 10 | 3605 | 100 | ||
| XM_008992873.1 | XP_008991121.1 | 8 - 9 | 2204 | 100 | ||
| XM_008992874.2 | XP_008991122.1 | 9 - 10 | 1503 | 100 | ||
| XM_008992875.1 | XP_008991123.1 | 8 - 9 | 1360 | 100 | ||
More Information
Set Membership: |
|

LEF1
Genomic Map