Assay Details
| Gene Symbol: NPAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753796.4 | XP_002753842.2 | 11 - 12 | 1521 | 82 | |
| XM_009005915.2 | XP_009004163.1 | 11 - 12 | 1502 | 82 | ||
More Information
Set Membership: |
|

NPAS3
Genomic Map