Assay Details
| Gene Symbol: CRLF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967473.1 | XP_017822962.1 | 4 - 5 | 873 | 102 | |
| XM_017967474.1 | XP_017822963.1 | 4 - 5 | 873 | 102 | ||
More Information
Set Membership: |
|

CRLF1
Genomic Map