Assay Details
| Gene Symbol: ARHGEF33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980806.2 | XP_008979054.1 | 14 - 15 | 2932 | 91 | |
| XM_008980808.2 | XP_008979056.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980809.2 | XP_008979057.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980810.2 | XP_008979058.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980811.2 | XP_008979059.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980812.2 | XP_008979060.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980813.2 | XP_008979061.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980814.2 | XP_008979062.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980816.2 | XP_008979064.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980817.2 | XP_008979065.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980819.2 | XP_008979067.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980820.2 | XP_008979068.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980821.2 | XP_008979069.1 | 14 - 15 | 2932 | 91 | ||
| XM_008980822.2 | XP_008979070.1 | 14 - 15 | 2932 | 91 | ||
| XM_017964394.1 | XP_017819883.1 | 14 - 15 | 2932 | 91 | ||
More Information
Set Membership: |
|

ARHGEF33
Genomic Map