Assay Details
| Gene Symbol: DUSP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979306.2 | XP_008977554.1 | 2 - 3 | 729 | 85 | |
| XM_008979307.1 | XP_008977555.1 | 2 - 3 | 726 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUSP26| Gene Symbol: DUSP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979306.2 | XP_008977554.1 | 2 - 3 | 729 | 85 | |
| XM_008979307.1 | XP_008977555.1 | 2 - 3 | 726 | 85 | ||