Assay Details
Gene Symbol: COMMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002757715.3 | XP_002757761.1 | 2 - 3 | 528 | 103 | |
XM_017964286.1 | XP_017819775.1 | 3 - 4 | 543 | 103 | ||
XM_017964291.1 | XP_017819780.1 | 3 - 4 | 351 | 103 | ||
XM_017964292.1 | XP_017819781.1 | 4 - 5 | 595 | 103 | ||
XM_017964293.1 | XP_017819782.1 | 2 - 3 | 384 | 103 | ||
XM_017964294.1 | XP_017819783.1 | 3 - 4 | 562 | 103 | ||
XM_017964295.1 | XP_017819784.1 | 2 - 3 | 310 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
