Assay Details
| Gene Symbol: C12H16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755881.2 | XP_002755927.1 | 2 - 3 | 1028 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12H16orf72| Gene Symbol: C12H16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755881.2 | XP_002755927.1 | 2 - 3 | 1028 | 81 | |