Assay Details
| Gene Symbol: TMEM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986225.2 | XP_008984473.1 | 2 - 3 | 901 | 65 | |
| XM_017966794.1 | XP_017822283.1 | 2 - 3 | 1155 | 65 | ||
| XM_017966795.1 | XP_017822284.1 | 2 - 3 | 1150 | 65 | ||
| XM_017966796.1 | XP_017822285.1 | 3 - 4 | 662 | 65 | ||
More Information
Set Membership: |
|

TMEM170A
Genomic Map