Assay Details
| Gene Symbol: ATRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981594.2 | XP_008979842.1 | 11 - 12 | 2159 | 75 | |
| XM_017964883.1 | XP_017820372.1 | 11 - 12 | 2156 | 75 | ||
More Information
Set Membership: |
|

ATRIP
Genomic Map