Assay Details
| Gene Symbol: UBE2W | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759002.4 | XP_002759048.1 | 4 - 5 | 461 | 76 | |
| XM_002759003.4 | XP_002759049.2 | 5 - 6 | 493 | 76 | ||
| XM_008982789.2 | XP_008981037.2 | 4 - 5 | 874 | 76 | ||
| XM_008982790.2 | XP_008981038.1 | 4 - 5 | 958 | 76 | ||
More Information
Set Membership: |
|

UBE2W
Genomic Map