Assay Details
| Gene Symbol: YWHAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747823.4 | XP_002747869.1 | 4 - 5 | 828 | 122 | |
| XM_008996385.2 | XP_008994633.1 | 4 - 5 | 715 | 122 | ||
| XM_008996386.1 | XP_008994634.1 | 5 - 6 | 762 | 122 | ||
More Information
Set Membership: |
|

YWHAE
Genomic Map