Assay Details
| Gene Symbol: PRRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762768.3 | XP_002762814.1 | 3 - 4 | 337 | 87 | |
| XM_008989136.2 | XP_008987384.1 | 3 - 4 | 1146 | 87 | ||
More Information
Set Membership: |
|

PRRG1
Genomic Map