Assay Details
| Gene Symbol: LOC100387916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758966.3 | XP_002759012.1 | 2 - 3 | 1131 | 86 | |
| XM_008982729.2 | XP_008980977.1 | 3 - 4 | 667 | 86 | ||
| XM_008982730.2 | XP_008980978.1 | 2 - 3 | 322 | 86 | ||
| XM_008982735.2 | XP_008980983.1 | 3 - 4 | 667 | 86 | ||
| XM_008982736.2 | XP_008980984.1 | 2 - 3 | 322 | 86 | ||
| XM_017965376.1 | XP_017820865.1 | 3 - 4 | 381 | 86 | ||
| XM_017965377.1 | XP_017820866.1 | 2 - 3 | 310 | 86 | ||
| XM_017965378.1 | XP_017820867.1 | 2 - 3 | 372 | 86 | ||
More Information
Set Membership: |
|

LOC100387916
Genomic Map