Assay Details
| Gene Symbol: ANXA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759895.4 | XP_002759941.1 | 14 - 15 | 1349 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANXA9| Gene Symbol: ANXA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759895.4 | XP_002759941.1 | 14 - 15 | 1349 | 73 | |
Set Membership: |
|