Assay Details
| Gene Symbol: PPP4R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734878.3 | XP_003734926.1 | 5 - 6 | 673 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP4R2| Gene Symbol: PPP4R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734878.3 | XP_003734926.1 | 5 - 6 | 673 | 91 | |
Set Membership: |
|