Assay Details
| Gene Symbol: ADCYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757102.2 | XP_002757148.1 | 5 - 6 | 836 | 102 | |
| XM_008979573.2 | XP_008977821.1 | 4 - 5 | 1021 | 102 | ||
| XM_008979574.2 | XP_008977822.1 | 3 - 4 | 609 | 102 | ||
| XM_017963908.1 | XP_017819397.1 | 3 - 4 | 453 | 102 | ||
More Information
Set Membership: |
|

ADCYAP1
Genomic Map