Assay Details
| Gene Symbol: XCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760410.4 | XP_002760456.1 | 2 - 3 | 930 | 90 | |
| XM_008985066.2 | XP_008983314.1 | 3 - 4 | 818 | 90 | ||
More Information
Set Membership: |
|

XCL1
Genomic Map