Assay Details
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003733406.3 | XP_003733454.2 | 2 - 3 | 532 | 71 | |
XM_017974579.1 | XP_017830068.1 | 2 - 3 | 519 | 71 | ||
XM_017974580.1 | XP_017830069.1 | 2 - 3 | 992 | 71 | ||
XM_017974581.1 | XP_017830070.1 | 2 - 3 | 527 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
