Assay Details
| Gene Symbol: SDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992291.2 | XP_008990539.1 | 8 - 9 | 1179 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SDHA| Gene Symbol: SDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992291.2 | XP_008990539.1 | 8 - 9 | 1179 | 75 | |
Set Membership: |
|