Assay Details
| Gene Symbol: PMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749560.4 | XP_002749606.1 | 7 - 8 | 996 | 93 | |
| XM_008999078.2 | XP_008997326.1 | 4 - 5 | 765 | 93 | ||
| XM_017973568.1 | XP_017829057.1 | 6 - 7 | 1153 | 93 | ||
| XM_017973569.1 | XP_017829058.1 | 6 - 7 | 1155 | 93 | ||
| XM_017973570.1 | XP_017829059.1 | 6 - 7 | 1157 | 93 | ||
More Information
Set Membership: |
|

PMS1
Genomic Map