Assay Details
| Gene Symbol: WDR20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754293.2 | XP_002754339.2 | 1 - 2 | 356 | 103 | |
| XM_009006569.1 | XP_009004817.1 | 1 - 2 | 356 | 103 | ||
| XM_009006570.1 | XP_009004818.1 | 1 - 2 | 356 | 103 | ||
| XM_009006573.2 | XP_009004821.1 | 1 - 2 | 356 | 103 | ||
| XM_017977336.1 | XP_017832825.1 | 1 - 2 | 356 | 103 | ||
| XM_017977337.1 | XP_017832826.1 | 1 - 2 | 356 | 103 | ||
More Information
Set Membership: |
|

WDR20
Genomic Map