Assay Details
| Gene Symbol: ZBTB49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745995.3 | XP_002746041.1 | 6 - 7 | 1639 | 98 | |
| XM_017970675.1 | XP_017826164.1 | 5 - 6 | 2741 | 98 | ||
More Information
Set Membership: |
|

ZBTB49
Genomic Map